Gene Transcripts
- Gene Symbol
- ACTN3
- Entrez Gene ID
- 89
- Gene Name
- actinin alpha 3 (gene/pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 11: 66546395 - 66563329 on Build GRCh38
- UniGene ID
- Hs.654432
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001104.3 |
NP_001095.2
|
15, 16 |
1951 |
|
NM_001258371.2 |
NP_001245300.2
|
15, 16 |
2099 |
|
NR_047663.2 |
|
15, 16 |
1951 |
|
GenBank mRNA |
AK125851.1 |
- |
15, 16 |
2485 |
|
AK303044.1 |
- |
15, 16 |
2099 |
|
AK313556.1 |
- |
15, 16 |
1889 |
|
BC096323.2 |
- |
9, 10 |
1011 |
|
BC099647.3 |
- |
15, 16 |
1872 |
40008220 |
BC099648.1 |
- |
5, 6 |
620 |
|
BC099649.3 |
- |
15, 16 |
1872 |
40008223 |
DA827389.1 |
- |
4, 5 |
535 |
|
M86407.1 |
- |
15, 16 |
1872 |
|
View Full Product Details