Gene Transcripts

Gene Symbol
AMBP
Entrez Gene ID
259
Gene Name
alpha-1-microglobulin/bikunin precursor
Gene Aliases
A1M, EDC1, HCP, HI30, IATIL, ITI, ITIL, ITILC, UTI
Chromosome Location
Chr. 9: 114060127 - 114078472 on Build GRCh38
UniGene ID
Hs.436911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001633.3 NP_001624.1 8 1024
GenBank mRNA AK290837.1 - 8 852
AK303795.1 - 8 750
AK310791.1 - 8 1041
AY544123.1 - 8 821
BC041593.1 - 8 987 5433460
X04225.1 - 8 833
X04494.1 - 8 806

View Full Product Details