Gene Transcripts

Gene Symbol
RND3
Entrez Gene ID
390
Gene Name
Rho family GTPase 3
Gene Aliases
ARHE, Rho8, RhoE, memB
Chromosome Location
Chr. 2: 150468193 - 150487695 on Build GRCh38
UniGene ID
Hs.6838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001254738.1 NP_001241667.1 3 564
NM_005168.4 NP_005159.1 4 471
GenBank mRNA AF498969.1 - 3 286
AK298007.1 - 4 441
AK299731.1 - 3 271
AK309442.1 - 2 145
AK313224.1 - 3 538
AL702945.1 - 3 537
BC012513.1 - 4 426 4453777
BQ440721.1 - 3 540
BT006769.1 - 3 286
CR542038.1 - 3 241
CR542070.1 - 3 241
DC367501.1 - 4 472
S82240.1 - 3 265
X95282.1 - 3 286
X97758.1 - 3 508

View Full Product Details