Gene Transcripts

Gene Symbol
KLF9
Entrez Gene ID
687
Gene Name
Kruppel like factor 9
Gene Aliases
BTEB, BTEB1
Chromosome Location
Chr. 9: 70384597 - 70414657 on Build GRCh38
UniGene ID
Hs.150557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206.2 NP_001197.1 1 1498
GenBank mRNA AB451404.1 - 1 204
AK313631.1 - 1 279
BC069431.1 - 1 277 7262239
BC074879.2 - 1 253 30915238
BC074880.2 - 1 253 30915472
D31716.1 - 1 1468

View Full Product Details