Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000732.4 | NP_000723.1 | 2 | 236 | |
NM_001040651.1 | NP_001035741.1 | 2 | 236 | ||
XM_017018543.1 | XP_016874032.1 | 3 | 1136 | ||
GenBank mRNA | AK311971.1 | - | 2 | 129 | |
BC039035.1 | - | 2 | 100 | 5745355 | |
BC070321.1 | - | 2 | 236 | 30412345 | |
CD014058.1 | - | 2 | 123 |