Gene Transcripts
- Gene Symbol
- CRYBB3
- Entrez Gene ID
- 1417
- Gene Name
- crystallin beta B3
- Gene Aliases
- CATCN2, CRYB3, CTRCT22
- Chromosome Location
- Chr. 22: 25199312 - 25207617 on Build GRCh38
- UniGene ID
- Hs.533022
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004076.4 |
NP_004067.1
|
2, 3 |
155 |
|
XM_011529902.2 |
XP_011528204.1
|
2, 3 |
693 |
|
XM_017028599.1 |
XP_016884088.1
|
2, 3 |
693 |
|
GenBank mRNA |
BC069479.1 |
- |
2, 3 |
125 |
|
BC102021.1 |
- |
2, 3 |
112 |
40029651 |
BC102022.1 |
- |
2, 3 |
112 |
40029650 |
BC107482.1 |
- |
2, 3 |
112 |
40029652 |
BM711382.1 |
- |
2, 3 |
155 |
|
CR456427.1 |
- |
1, 2 |
106 |
|
U71216.1 |
- |
2, 3 |
146 |
|
View Full Product Details