Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000774.4 | NP_000765.2 | 9 | 1236 | |
NR_135528.1 | 9 | 1178 | |||
XM_011526551.2 | XP_011524853.1 | 9 | 1376 | ||
XM_011526552.2 | XP_011524854.1 | 9 | 1379 | ||
XM_011526554.2 | XP_011524856.1 | 8 | 1194 | ||
XM_017026384.1 | XP_016881873.1 | 8 | 1567 | ||
XM_017026385.1 | XP_016881874.1 | 8 | 1567 | ||
GenBank mRNA | AK298082.1 | - | 8 | 1767 | |
AK316498.1 | - | 9 | 1178 | ||
BC109056.1 | - | 9 | 1155 | 40033365 | |
J02906.1 | - | 9 | 1219 |