Gene Transcripts

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
Chromosome Location
Chr. 5: 13690328 - 14011829 on Build GRCh38
UniGene ID
Hs.212360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001369.2 NP_001360.1 14 1842
XM_005248262.3 XP_005248319.2 14 1925
XM_017009177.1 XP_016864666.1 14 1925
XM_017009178.1 XP_016864667.1 14 2728
XM_017009179.1 XP_016864668.1 8 1022
XM_017009180.1 XP_016864669.1 14 1925
XM_017009181.1 XP_016864670.1 14 1925
XM_017009182.1 XP_016864671.1 14 1925
XM_017009183.1 XP_016864672.1 14 1925
XM_017009184.1 XP_016864673.1 14 1925
XM_017009187.1 XP_016864676.1 14 1925
GenBank mRNA AY045575.1 - 14 1842

View Full Product Details