Gene Transcripts

Gene Symbol
DOK1
Entrez Gene ID
1796
Gene Name
docking protein 1
Gene Aliases
P62DOK, pp62
Chromosome Location
Chr. 2: 74549020 - 74557551 on Build GRCh38
UniGene ID
Hs.103854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197260.1 NP_001184189.1 5 1097
NM_001318866.1 NP_001305795.1 5 1597
NM_001318867.1 NP_001305796.1 5 1642
NM_001318868.1 NP_001305797.1 4 1414
NM_001318869.1 NP_001305798.1 4 1408
NM_001381.4 NP_001372.1 5 1599
XM_017003528.1 XP_016859017.1 3 1520
GenBank mRNA AF035299.1 - 5 1090
AF180527.1 - 4 819
AK055944.1 - 5 1599
AK127127.1 - 3 1295
AK127656.1 - 3 1295
AK222883.1 - 5 1038
AK296154.1 - 5 1052
AK300097.1 - 4 819
AK314678.1 - 5 1019
BC111055.1 - 4 843 5752188
BC114440.1 - 5 1025 40083624
BC114497.1 - 4 828 40083623
U70987.1 - 5 1026

View Full Product Details