Gene Transcripts
- Gene Symbol
- DOK1
- Entrez Gene ID
- 1796
- Gene Name
- docking protein 1
- Gene Aliases
- P62DOK, pp62
- Chromosome Location
- Chr. 2: 74549020 - 74557551 on Build GRCh38
- UniGene ID
- Hs.103854
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001197260.1 |
NP_001184189.1
|
3, 4 |
551 |
|
NM_001318866.1 |
NP_001305795.1
|
3, 4 |
1051 |
|
NM_001318867.1 |
NP_001305796.1
|
3, 4 |
1096 |
|
NM_001381.4 |
NP_001372.1
|
3, 4 |
1053 |
|
GenBank mRNA |
AF035299.1 |
- |
3, 4 |
544 |
|
AK055944.1 |
- |
3, 4 |
1053 |
|
AK222883.1 |
- |
3, 4 |
492 |
|
AK296154.1 |
- |
3, 4 |
506 |
|
AK314678.1 |
- |
3, 4 |
473 |
|
BC114440.1 |
- |
3, 4 |
479 |
40083624 |
DA382785.1 |
- |
3, 4 |
551 |
|
U70987.1 |
- |
3, 4 |
480 |
|
View Full Product Details