Gene Transcripts

Gene Symbol
DSG1
Entrez Gene ID
1828
Gene Name
desmoglein 1
Gene Aliases
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1
Chromosome Location
Chr. 18: 31318089 - 31359246 on Build GRCh38
UniGene ID
Hs.2633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001942.3 NP_001933.2 15 3017
GenBank mRNA AF097935.1 - 15 3018
AK302888.1 - 4 1191
X56654.1 - 15 2882

View Full Product Details