Gene Transcripts

Gene Symbol
EDN1
Entrez Gene ID
1906
Gene Name
endothelin 1
Gene Aliases
ARCND3, ET1, HDLCQ7, PPET1, QME
Chromosome Location
Chr. 6: 12256464 - 12297194 on Build GRCh38
UniGene ID
Hs.511899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168319.1 NP_001161791.1 3 623
NM_001955.4 NP_001946.3 3 626
XM_011514330.2 XP_011512632.1 4 382
XM_011514331.2 XP_011512633.1 5 543
XM_011514332.2 XP_011512634.1 4 380
XM_017010331.1 XP_016865820.1 4 566
GenBank mRNA AK226096.1 - 3 541
AK291838.1 - 3 563
AK313914.1 - 3 558
BC009720.1 - 3 495 3900704
BX439515.2 - 3 586
CR541903.1 - 3 292
S56805.1 - 3 628
Y00749.1 - 3 544

View Full Product Details