Gene Transcripts
- Gene Symbol
- EYA2
- Entrez Gene ID
- 2139
- Gene Name
- EYA transcriptional coactivator and phosphatase 2
- Gene Aliases
- EAB1
- Chromosome Location
- Chr. 20: 46894624 - 47188844 on Build GRCh38
- UniGene ID
- Hs.472877
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005244.4 |
NP_005235.3
|
2, 3 |
480 |
|
NM_172110.3 |
NP_742108.2
|
2, 3 |
480 |
|
XM_005260327.2 |
XP_005260384.1
|
2, 3 |
173 |
|
XM_017027721.1 |
XP_016883210.1
|
2, 3 |
198 |
|
GenBank mRNA |
AK293032.1 |
- |
2, 3 |
184 |
|
AY705349.1 |
- |
1, 2 |
116 |
|
BC008803.2 |
- |
2, 3 |
234 |
3944547 |
BC013882.2 |
- |
2, 3 |
480 |
|
BT006682.1 |
- |
1, 2 |
106 |
|
U71207.1 |
- |
1, 2 |
108 |
|
View Full Product Details