Gene Transcripts

Gene Symbol
FABP7
Entrez Gene ID
2173
Gene Name
fatty acid binding protein 7
Gene Aliases
B-FABP, BLBP, FABPB, MRG
Chromosome Location
Chr. 6: 122749201 - 122784077 on Build GRCh38
UniGene ID
Hs.26770
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319039.1 NP_001305968.1 2 531
NM_001319041.1 NP_001305970.1 2 531
NM_001319042.1 NP_001305971.1 2 272
NM_001446.4 NP_001437.1 2 531
GenBank mRNA AB208815.1 - 2 278
AJ002962.1 - 2 287
AK289836.1 - 2 290
AK307272.1 - 2 291
AK311867.1 - 2 290
AL512688.1 - 2 247
AW156936.1 - 3 497
BC012299.1 - 2 397 4707233
BU736791.1 - 3 512
BX461253.2 - 2 252
CR457057.1 - 2 211
D50373.1 - 2 263
D88648.1 - 2 262
KM111186.1 - 2 272
U51338.1 - 2 266
U81235.1 - 2 226

View Full Product Details