Gene Transcripts

Gene Symbol
GALNS
Entrez Gene ID
2588
Gene Name
galactosamine (N-acetyl)-6-sulfatase
Gene Aliases
GALNAC6S, GAS, GalN6S, MPS4A
Chromosome Location
Chr. 16: 88813734 - 88856970 on Build GRCh38
UniGene ID
Hs.271383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000512.4 NP_000503.1 8 870
NM_001323543.1 NP_001310472.1 7 750
NM_001323544.1 NP_001310473.1 9 1044
XM_005256301.3 XP_005256358.1 8 870
XM_011522982.2 XP_011521284.1 9 916
XM_017023111.1 XP_016878600.1 8 2767
XM_017023112.1 XP_016878601.1 8 2767
XM_017023113.1 XP_016878602.1 7 728
GenBank mRNA AA436724.1 - 4 368
AK131096.1 - 6 4189
AK294390.1 - 6 620
AK308269.1 - 9 900
AK312655.1 - 8 847
AY129021.1 - 6 547
BC050684.1 - 8 859 6091623
BC056151.1 - 8 851 6026248
BX649181.1 - 6 463

View Full Product Details