Gene Transcripts

Gene Symbol
GPR17
Entrez Gene ID
2840
Gene Name
G protein-coupled receptor 17
Gene Aliases
Chromosome Location
Chr. 2: 127645864 - 127652639 on Build GRCh38
UniGene ID
Hs.46453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161415.1 NP_001154887.1 3, 4 686
NM_001161416.1 NP_001154888.1 2, 3 621
NM_001161417.1 NP_001154889.1 1, 2 392
NM_005291.2 NP_005282.1 2, 3 457
XM_017003833.1 XP_016859322.1 1, 2 813
GenBank mRNA AK126849.1 - 1 1647
AK292725.1 - 1, 2 262
AK313949.1 - 1, 2 148
BC031653.1 - 1, 2 71 5174146
BC039595.1 - 1, 2 76
BX538082.1 - 1, 2 206
DA154324.1 - 1, 2 392
DA314295.1 - 2, 3 169
DA366480.1 - 2, 3 493
DA390892.1 - 1, 2 371
Y12546.1 - 2, 3 149
Z94154.1 - 1, 2 85

View Full Product Details