Gene Transcripts

Gene Symbol
HOXC11
Entrez Gene ID
3227
Gene Name
homeobox C11
Gene Aliases
HOX3H
Chromosome Location
Chr. 12: 53973126 - 53976419 on Build GRCh38
UniGene ID
Hs.127562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014212.3 NP_055027.1 1 450
GenBank mRNA AJ000041.1 - 1 378
AK291946.1 - 1 448
BC001543.1 - 1 431 3462682

View Full Product Details