Gene Transcripts
- Gene Symbol
- INPP5A
- Entrez Gene ID
- 3632
- Gene Name
- inositol polyphosphate-5-phosphatase A
- Gene Aliases
- 5PTASE
- Chromosome Location
- Chr. 10: 132537779 - 132783480 on Build GRCh38
- UniGene ID
- Hs.523360
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321042.1 |
NP_001307971.1
|
5, 6 |
1011 |
|
NM_005539.4 |
NP_005530.3
|
8, 9 |
958 |
|
XM_017016204.1 |
XP_016871693.1
|
7, 8 |
612 |
|
XM_017016205.1 |
XP_016871694.1
|
8, 9 |
953 |
|
GenBank mRNA |
AF273055.1 |
- |
8, 9 |
901 |
|
BC052265.1 |
- |
7, 8 |
556 |
|
BC062300.1 |
- |
7, 8 |
556 |
|
BC096280.2 |
- |
9, 10 |
702 |
40006844 |
BC096281.2 |
- |
8, 9 |
621 |
40006846 |
X77567.1 |
- |
8, 9 |
739 |
|
Z31695.1 |
- |
8, 9 |
594 |
|
View Full Product Details