Gene Transcripts

Gene Symbol
KPNA3
Entrez Gene ID
3839
Gene Name
karyopherin subunit alpha 3
Gene Aliases
IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
Chromosome Location
Chr. 13: 49699307 - 49792921 on Build GRCh38
UniGene ID
Hs.527919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002267.3 NP_002258.2 17 1923
XM_017020561.1 XP_016876050.1 17 1606
GenBank mRNA AF005263.1 - 17 1520
AF034756.1 - 17 1510
AK223480.1 - 17 1545
AK290528.1 - 17 1583
AK291000.1 - 17 1547
BC017355.2 - 17 1532 5020312
BC024202.2 - 17 1532 3531069
BC035090.2 - 17 1924 5261465
D89618.1 - 17 1599
Y12394.1 - 17 1523

View Full Product Details