Gene Transcripts

Gene Symbol
MAB21L1
Entrez Gene ID
4081
Gene Name
mab-21 like 1
Gene Aliases
CAGR1, Nbla00126
Chromosome Location
Chr. 13: 35473789 - 35476695 on Build GRCh38
UniGene ID
Hs.584776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005584.4 NP_005575.1 1 893
GenBank mRNA AB073388.1 - 1 883
AB593021.1 - 1 814
AB593022.1 - 1 884
AK313185.1 - 1 869
BC028170.1 - 1 814 5244028
CR457420.1 - 1 336

View Full Product Details