Gene Transcripts

Gene Symbol
PDC
Entrez Gene ID
5132
Gene Name
phosducin
Gene Aliases
MEKA, PHD, PhLOP, PhLP
Chromosome Location
Chr. 1: 186443566 - 186461108 on Build GRCh38
UniGene ID
Hs.654381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002597.4 NP_002588.3 2, 3 145
NM_022576.3 NP_072098.1 1, 2 107
XM_011509603.2 XP_011507905.1 1, 2 1163
GenBank mRNA AF076464.1 - 1, 2 108
BQ636049.1 - 2, 3 144
EL945964.1 - 2, 3 149
M33478.1 - 2, 3 122

View Full Product Details