Gene Transcripts

Gene Symbol
PGR
Entrez Gene ID
5241
Gene Name
progesterone receptor
Gene Aliases
NR3C3, PR
Chromosome Location
Chr. 11: 101029624 - 101130681 on Build GRCh38
UniGene ID
Hs.32405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000926.4 NP_000917.3 1 1676
NM_001202474.3 NP_001189403.1 1 926
NM_001271161.2 NP_001258090.1 1 926
NR_073141.2 1 926
NR_073142.2 1 926
NR_073143.2 1 926
XM_006718858.3 XP_006718921.1 1 2526
XM_011542869.2 XP_011541171.1 1 2544
GenBank mRNA AB084248.1 - 1 933
AB085683.1 - 1 933
AB085843.1 - 1 933
AB085844.1 - 1 933
AB085845.1 - 1 933
AF016381.1 - 1 1108
M15716.1 - 1 1108
X51730.1 - 1 2387

View Full Product Details