Gene Transcripts
- Gene Symbol
- PPP4C
- Entrez Gene ID
- 5531
- Gene Name
- protein phosphatase 4 catalytic subunit
- Gene Aliases
- PP-X, PP4, PP4C, PPH3, PPP4, PPX
- Chromosome Location
- Chr. 16: 30075976 - 30085377 on Build GRCh38
- UniGene ID
- Hs.534338
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303503.1 |
NP_001290432.1
|
6, 7 |
658 |
|
NM_001303504.1 |
NP_001290433.1
|
5, 6 |
550 |
|
NM_001303506.1 |
NP_001290435.1
|
6, 7 |
711 |
|
NM_001303507.1 |
NP_001290436.1
|
5, 6 |
600 |
|
NM_002720.2 |
NP_002711.1
|
6, 7 |
652 |
|
XM_006721061.3 |
XP_006721124.1
|
5, 6 |
537 |
|
XM_017023403.1 |
XP_016878892.1
|
6, 7 |
628 |
|
XM_017023404.1 |
XP_016878893.1
|
5, 6 |
564 |
|
GenBank mRNA |
AF097996.1 |
- |
6, 7 |
574 |
|
BC001416.2 |
- |
6, 7 |
605 |
3140324 |
BG706310.1 |
- |
5, 6 |
604 |
|
BG741669.1 |
- |
6, 7 |
684 |
|
BQ220878.1 |
- |
5, 6 |
460 |
|
BT019746.1 |
- |
5, 6 |
470 |
|
X70218.1 |
- |
6, 7 |
605 |
|
View Full Product Details