Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199163.1 | NP_001186092.1 | 7 | 862 | |
NM_002805.5 | NP_002796.4 | 7 | 727 | ||
GenBank mRNA | AF035309.1 | - | 805 | ||
AK095836.1 | - | 6 | 1961 | ||
AK098484.1 | - | 1891 | |||
BC001932.1 | - | 7 | 678 | 2820259 | |
BC002367.2 | - | 7 | 657 | 2821416 | |
BC127888.1 | - | 5 | 569 | ||
D44467.1 | - | 7 | 657 |