Gene Transcripts

Gene Symbol
RFX5
Entrez Gene ID
5993
Gene Name
regulatory factor X5
Gene Aliases
Chromosome Location
Chr. 1: 151340640 - 151347319 on Build GRCh38
UniGene ID
Hs.632472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000449.3 NP_000440.1 11 1878
NM_001025603.1 NP_001020774.1 11 1871
XM_005245405.1 XP_005245462.1 11 1933
XM_005245406.3 XP_005245463.1 11 1890
XM_011509847.1 XP_011508149.1 11 1937
XM_011509848.1 XP_011508150.1 11 1940
XM_011509849.1 XP_011508151.1 11 1873
XM_011509850.1 XP_011508152.1 11 2000
XM_017001999.1 XP_016857488.1 11 2001
XM_017002000.1 XP_016857489.1 11 2032
GenBank mRNA AK223526.1 - 9 1670
AK302891.1 - 10 1751
AK312289.1 - 11 1851
AL050135.1 - 3 1113
BC017471.1 - 11 1805 3909092
X85786.1 - 10 1818

View Full Product Details