Gene Transcripts

Gene Symbol
TNNT3
Entrez Gene ID
7140
Gene Name
troponin T3, fast skeletal type
Gene Aliases
TNTF
Chromosome Location
Chr. 11: 1919551 - 1938706 on Build GRCh38
UniGene ID
Hs.73454
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042780.2 NP_001036245.1 13 825
NM_001042781.2 NP_001036246.1 14 843
NM_001042782.2 NP_001036247.1 13 825
NM_001297646.1 NP_001284575.1 14 1083
NM_006757.3 NP_006748.1 14 849
XM_006718288.3 XP_006718351.1 9 876
XM_006718290.3 XP_006718353.1 16 788
XM_006718293.2 XP_006718356.1 15 865
XM_006718294.3 XP_006718357.1 15 865
XM_006718296.3 XP_006718359.1 14 752
XM_006718299.2 XP_006718362.1 13 688
XM_006718300.3 XP_006718363.1 12 716
XM_006718302.3 XP_006718365.1 11 698
XM_011520343.2 XP_011518645.1 16 919
XM_017018205.1 XP_016873694.1 14 843
XM_017018206.1 XP_016873695.1 16 711
XM_017018207.1 XP_016873696.1 15 699
XM_017018208.1 XP_016873697.1 14 666
XM_017018209.1 XP_016873698.1 14 675
XM_017018210.1 XP_016873699.1 13 640
XM_017018211.1 XP_016873700.1 13 659
XM_017018212.1 XP_016873701.1 12 631
XM_017018213.1 XP_016873702.1 11 616
XM_017018214.1 XP_016873703.1 11 615
XM_017018215.1 XP_016873704.1 11 607
XM_017018216.1 XP_016873705.1 15 770
XM_017018217.1 XP_016873706.1 14 749
XM_017018218.1 XP_016873707.1 13 737
XM_017018219.1 XP_016873708.1 11 698
GenBank mRNA AI161366.1 - 3 353
AK056968.1 - 13 904
BC050446.1 - 13 681
BC117327.1 - 12 709 40125878
BC143537.1 - 12 709 9052047
BC171727.1 - 13 727 9052046
BC171728.1 - 9 662
BT019997.1 - 13 631
BX640689.1 - 14 707
CR541927.1 - 13 631
DQ778624.1 - 9 613
M21984.1 - 13 649

View Full Product Details