Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001326578.1 | NP_001313507.1 | 9 | 1003 | |
NM_001326579.1 | NP_001313508.1 | 9 | 790 | ||
NM_001326580.1 | NP_001313509.1 | 9 | 926 | ||
NM_003569.2 | NP_003560.2 | 9 | 926 | ||
NR_137169.1 | 8 | 853 | |||
XM_011536178.1 | XP_011534480.1 | 9 | 842 | ||
GenBank mRNA | AK294911.1 | - | 9 | 675 | |
AK301464.1 | - | 3 | 289 | ||
BC011975.1 | - | 9 | 790 | 3860059 | |
U77942.1 | - | 9 | 755 |