Gene Transcripts

Gene Symbol
TPST1
Entrez Gene ID
8460
Gene Name
tyrosylprotein sulfotransferase 1
Gene Aliases
TANGO13A
Chromosome Location
Chr. 7: 66205272 - 66360451 on Build GRCh38
UniGene ID
Hs.421194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003596.3 NP_003587.1 2 695
XM_005250642.2 XP_005250699.1 2 691
XM_011516634.2 XP_011514936.1 2 691
XM_011516635.2 XP_011514937.1 2 691
XM_017012724.1 XP_016868213.1 3 778
XM_017012725.1 XP_016868214.1 3 778
XM_017012726.1 XP_016868215.1 2 691
XM_017012727.1 XP_016868216.1 2 691
GenBank mRNA AF038009.1 - 1 424
AK313098.1 - 2 633
BC013188.1 - 2 670 3918922
CR542060.1 - 1 343
CR542080.1 - 1 343

View Full Product Details