Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252641.1 | NP_001239570.1 | 5 | 423 | |
NM_003796.3 | NP_003787.2 | 5 | 738 | ||
NR_045557.1 | 4 | 703 | |||
XM_005259362.2 | XP_005259419.1 | 5 | 417 | ||
XM_005259363.4 | XP_005259420.1 | 5 | 474 | ||
XM_011527435.1 | XP_011525737.1 | 4 | 300 | ||
GenBank mRNA | AF091095.1 | - | 6 | 374 | |
AK001437.1 | - | 4 | 424 | ||
AK226173.1 | - | 5 | 726 | ||
AK292170.1 | - | 5 | 474 | ||
AK314313.1 | - | 5 | 463 | ||
BC026184.2 | - | 5 | 423 | ||
BC067259.1 | - | 4 | 430 | 6500727 |