Gene Transcripts
- Gene Symbol
- MYH13
- Entrez Gene ID
- 8735
- Gene Name
- myosin heavy chain 13
- Gene Aliases
- MyHC-IIL, MyHC-eo
- Chromosome Location
- Chr. 17: 10300866 - 10373005 on Build GRCh38
- UniGene ID
- Hs.711142
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003802.2 |
NP_003793.2
|
23 |
2816 |
|
GenBank mRNA |
AF111782.2 |
- |
23 |
2816 |
|
View Full Product Details