Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003842.4 | NP_003833.4 | 9 | 1439 | |
NM_147187.2 | NP_671716.2 | 10 | 1352 | ||
NR_027140.1 | 9 | 1246 | |||
XM_017013944.1 | XP_016869433.1 | 9 | 1227 | ||
GenBank mRNA | AB451272.1 | - | 9 | 1146 | |
AB451399.1 | - | 9 | 1146 | ||
AF012535.1 | - | 10 | 1198 | ||
AF012628.1 | - | 10 | 1059 | ||
AF016266.1 | - | 9 | 1263 | ||
AF016268.1 | - | 10 | 1168 | ||
AF016849.1 | - | 9 | 1290 | ||
AF018657.1 | - | 10 | 1059 | ||
AF018658.1 | - | 9 | 1146 | ||
AF020501.1 | - | 10 | 1059 | ||
AF022386.1 | - | 10 | 1059 | ||
AF153687.1 | - | 10 | 1177 | ||
AF192548.1 | - | 10 | 1159 | ||
AK291216.1 | - | 9 | 1266 | ||
AK296128.1 | - | 5 | 452 | ||
AK298602.1 | - | 9 | 1090 | ||
AY358277.1 | - | 10 | 1198 | ||
BC001281.1 | - | 9 | 1293 | 3458466 | |
BX537617.1 | - | 4 | 544 | ||
BX538104.1 | - | 9 | 1268 | ||
BX538179.1 | - | 4 | 544 | ||
CR541898.1 | - | 9 | 1146 | ||
CX870630.1 | - | 5 | 651 | ||
FJ200482.1 | - | 8 | 1040 |