Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282285.1 | NP_001269214.1 | 5 | 651 | |
NM_001282286.1 | NP_001269215.1 | 5 | 648 | ||
NM_003845.2 | NP_003836.1 | 13 | 1547 | ||
GenBank mRNA | AA524246.1 | - | 1 | 320 | |
AF263541.1 | - | 12 | 1444 | ||
AK292322.1 | - | 13 | 1541 | ||
AK298979.1 | - | 4 | 468 | ||
AK303744.1 | - | 5 | 616 | ||
AL561586.3 | - | 5 | 648 | ||
BC031244.1 | - | 13 | 1541 | 5266287 | |
GU139739.1 | - | 3 | 355 | ||
GU139740.1 | - | 4 | 522 | ||
GU139741.1 | - | 5 | 583 | ||
GU139742.1 | - | 5 | 564 | ||
GU139743.1 | - | 12 | 1531 | ||
GU139744.1 | - | 4 | 518 | ||
GU139745.1 | - | 1217 | |||
HY337672.1 | - | 1 | 342 | ||
Y09305.1 | - | 5 | 542 |