Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
8800
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
PEX11-ALPHA, PMP28, hsPEX11p
Chromosome Location
Chr. 15: 89681531 - 89690784 on Build GRCh38
UniGene ID
Hs.31034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271572.1 NP_001258501.1 2 243
NM_003847.2 NP_003838.1 2 243
GenBank mRNA AB015594.1 - 2 186
AF093668.1 - 2 148
AK001415.1 - 2 236
AK300978.1 - 2 121
AK313318.1 - 2 157
BC009697.1 - 2 137 3878406
BC101164.2 - 2 97 40015908
BC114384.1 - 2 97 40004153
CR542046.1 - 2 91
CR542075.1 - 2 91

View Full Product Details