Gene Transcripts
- Gene Symbol
- NPEPPS
- Entrez Gene ID
- 9520
- Gene Name
- aminopeptidase puromycin sensitive
- Gene Aliases
- AAP-S, MP100, PSA
- Chromosome Location
- Chr. 17: 47522950 - 47623276 on Build GRCh38
- UniGene ID
- Hs.443837
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006310.3 |
NP_006301.3
|
17, 18 |
2314 |
|
XM_011525496.1 |
XP_011523798.1
|
18, 19 |
2116 |
|
XM_017025372.1 |
XP_016880861.1
|
17, 18 |
2276 |
|
XM_017025373.1 |
XP_016880862.1
|
18, 19 |
2116 |
|
GenBank mRNA |
AJ132583.1 |
- |
17, 18 |
2044 |
|
AK096491.1 |
- |
2, 3 |
772 |
|
AK096709.1 |
- |
19, 20 |
2101 |
|
AK293450.1 |
- |
11, 12 |
1246 |
|
AK293995.1 |
- |
10, 11 |
1228 |
|
AK296887.1 |
- |
16, 17 |
1957 |
|
AK297107.1 |
- |
16, 17 |
1818 |
|
AK303037.1 |
- |
18, 19 |
2109 |
|
BC065294.1 |
- |
17, 18 |
2003 |
6059589 |
View Full Product Details