Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001178129.1 | NP_001171600.1 | 16 | 1804 | |
NM_012431.2 | NP_036563.1 | 16 | 2440 | ||
GenBank mRNA | AB002329.1 | - | 16 | 2285 | |
AK297209.1 | - | 16 | 2120 | ||
AK303925.1 | - | 16 | 1805 | ||
BC140706.1 | - | 16 | 1975 | 9021574 | |
BC144338.1 | - | 16 | 1975 | 9052858 | |
BC152458.1 | - | 16 | 2297 | 8862593 |