Gene Transcripts
- Gene Symbol
- SNX17
- Entrez Gene ID
- 9784
- Gene Name
- sorting nexin 17
- Gene Aliases
- Chromosome Location
- Chr. 2: 27370496 - 27377533 on Build GRCh38
- UniGene ID
- Hs.278569
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267059.1 |
NP_001253988.1
|
8, 9 |
885 |
|
NM_001267060.1 |
NP_001253989.1
|
7, 8 |
846 |
|
NM_001267061.1 |
NP_001253990.1
|
8, 9 |
756 |
|
NM_014748.3 |
NP_055563.1
|
8, 9 |
921 |
|
NR_049782.1 |
|
8, 9 |
1046 |
|
NR_049783.1 |
|
9, 10 |
1019 |
|
NR_049784.1 |
|
9, 10 |
995 |
|
NR_049785.1 |
|
7, 8 |
928 |
|
NR_049786.1 |
|
8, 9 |
877 |
|
NR_049787.1 |
|
6, 7 |
728 |
|
XM_011533203.1 |
XP_011531505.1
|
7, 8 |
695 |
|
XM_017005405.1 |
XP_016860894.1
|
6, 7 |
865 |
|
GenBank mRNA |
AK129689.1 |
- |
8, 9 |
805 |
|
AK222543.1 |
- |
8, 9 |
804 |
|
AK293252.1 |
- |
9, 10 |
895 |
|
AK293464.1 |
- |
8, 9 |
737 |
|
AK297054.1 |
- |
8, 9 |
930 |
|
AK298620.1 |
- |
8, 9 |
756 |
|
AK298856.1 |
- |
8, 9 |
758 |
|
AK298869.1 |
- |
7, 8 |
707 |
|
AK300144.1 |
- |
8, 9 |
768 |
|
AK300641.1 |
- |
6, 7 |
599 |
|
AK307122.1 |
- |
7, 8 |
804 |
|
AK308487.1 |
- |
7, 8 |
807 |
|
AK309640.1 |
- |
6, 7 |
849 |
|
AK316373.1 |
- |
8, 9 |
700 |
|
BC002524.1 |
- |
8, 9 |
782 |
3140610 |
BC002610.1 |
- |
8, 9 |
771 |
3143104 |
BC014620.2 |
- |
8, 9 |
782 |
3010648 |
BC050590.1 |
- |
8, 9 |
776 |
6044270 |
BT007167.1 |
- |
8, 9 |
673 |
|
CR457081.1 |
- |
8, 9 |
673 |
|
D31764.1 |
- |
8, 9 |
895 |
|
View Full Product Details