Gene Transcripts

Gene Symbol
FEM1B
Entrez Gene ID
10116
Gene Name
fem-1 homolog B
Gene Aliases
F1A-ALPHA, F1AA, FEM1-beta
Chromosome Location
Chr. 15: 68277803 - 68295865 on Build GRCh38
UniGene ID
Hs.362733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015322.4 NP_056137.1 2 1578
XM_017021859.1 XP_016877348.1 2 1091
GenBank mRNA AB007856.2 - 2 963
AF178632.1 - 2 963
AF204883.1 - 2 963
AK098620.1 - 1 99
AK290167.1 - 2 1312
AL833264.1 - 1 1910
BC010122.2 - 2 1578 3840453
BC014558.2 - 2 1578 3840377

View Full Product Details