Gene Transcripts

Gene Symbol
MPHOSPH9
Entrez Gene ID
10198
Gene Name
M-phase phosphoprotein 9
Gene Aliases
MPP-9, MPP9
Chromosome Location
Chr. 12: 123152324 - 123244014 on Build GRCh38
UniGene ID
Hs.577404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022782.3 NP_073619.3 18, 19 3078
NR_103517.1 18, 19 2817
XM_011537740.2 XP_011536042.1 18, 19 2982
XM_011537741.2 XP_011536043.1 18, 19 2982
XM_017018673.1 XP_016874162.1 18, 19 3076
XM_017018674.1 XP_016874163.1 17, 18 2880
XM_017018675.1 XP_016874164.1 17, 18 2884
XM_017018676.1 XP_016874165.1 17, 18 2826
XM_017018677.1 XP_016874166.1 18, 19 2982
XM_017018678.1 XP_016874167.1 17, 18 2802
XM_017018679.1 XP_016874168.1 19, 20 3015
XM_017018680.1 XP_016874169.1 17, 18 2924
XM_017018681.1 XP_016874170.1 17, 18 2837
GenBank mRNA AK023016.1 - 13, 14 1801
AK091181.1 - 14, 15 2407
AK096541.1 - 18, 19 2690
AL096751.1 - 12, 13 1738
BC130440.1 - 14, 15 2360 40146471
BC144016.1 - 13, 14 2234
BC144017.1 - 13, 14 2012

View Full Product Details