Gene Transcripts
- Gene Symbol
- PFDN6
- Entrez Gene ID
- 10471
- Gene Name
- prefoldin subunit 6
- Gene Aliases
- H2-KE2, HKE2, KE-2, PFD6
- Chromosome Location
- Chr. 6: 33289597 - 33290934 on Build GRCh38
- UniGene ID
- Hs.446374
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001185181.2 |
NP_001172110.1
|
1, 2 |
292 |
|
NM_001265595.1 |
NP_001252524.1
|
2, 3 |
184 |
|
NM_001265596.1 |
NP_001252525.1
|
2, 3 |
181 |
|
NM_014260.3 |
NP_055075.1
|
2, 3 |
189 |
|
GenBank mRNA |
AK312150.1 |
- |
2, 3 |
177 |
|
BC039033.1 |
- |
1, 2 |
158 |
5748799 |
BC059783.1 |
- |
1, 2 |
164 |
5415221 |
BE794658.1 |
- |
1, 2 |
145 |
|
BM686351.1 |
- |
1, 2 |
273 |
|
BM745352.1 |
- |
2, 3 |
184 |
|
BM792413.1 |
- |
1, 2 |
166 |
|
BM818422.1 |
- |
2, 3 |
184 |
|
BM834357.1 |
- |
2, 3 |
185 |
|
CB124036.1 |
- |
1, 2 |
292 |
|
DA256816.1 |
- |
2, 3 |
177 |
|
View Full Product Details