Gene Transcripts

Gene Symbol
PMVK
Entrez Gene ID
10654
Gene Name
phosphomevalonate kinase
Gene Aliases
HUMPMKI, PMK, PMKA, PMKASE, POROK1
Chromosome Location
Chr. 1: 154924732 - 154942650 on Build GRCh38
UniGene ID
Hs.30954
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323011.1 NP_001309940.1 3 186
NM_001323012.1 NP_001309941.1 3 193
NM_006556.3 NP_006547.1 3 536
GenBank mRNA AK130234.1 - 4 378
AK312803.1 - 3 495
BC006089.1 - 3 519 3604167
BC007694.2 - 3 449 3954909
BQ061044.1 - 3 536
BT019976.1 - 3 213
CR541998.1 - 3 213
L77213.1 - 3 248

View Full Product Details