Gene Transcripts

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
Chromosome Location
Chr. 14: 52857271 - 52951192 on Build GRCh38
UniGene ID
Hs.509343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134999.1 NP_001128471.1 15, 16 2086
NM_006832.2 NP_006823.1 14, 15 2065
XM_005267285.2 XP_005267342.1 15, 16 2193
XM_006720008.2 XP_006720071.1 17, 18 2238
XM_006720009.2 XP_006720072.1 16, 17 2214
XM_006720010.2 XP_006720073.1 16, 17 2205
GenBank mRNA AF443279.1 - 14, 15 1960
AK126426.1 - 2, 3 1115
AK291738.1 - 14, 15 2018
BC017327.2 - 14, 15 2065 4547604
CR749486.1 - 2 4748
EU979385.1 - 14, 15 1900
Z24725.1 - 14, 15 2001

View Full Product Details