Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007240.2 | NP_009171.1 | 4 | 667 | |
XM_005244862.2 | XP_005244919.1 | 4 | 756 | ||
GenBank mRNA | AF119226.1 | - | 4 | 644 | |
AK000259.1 | - | 2 | 81 | ||
BC006286.1 | - | 4 | 665 | 3958403 | |
BT006633.1 | - | 4 | 633 |