Gene Transcripts

Gene Symbol
MAPK8IP3
Entrez Gene ID
23162
Gene Name
mitogen-activated protein kinase 8 interacting protein 3
Gene Aliases
JIP-3, JIP3, JSAP1, SYD2, syd
Chromosome Location
Chr. 16: 1706183 - 1770317 on Build GRCh38
UniGene ID
Hs.207763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040439.1 NP_001035529.1 6 1096
NM_001318852.1 NP_001305781.1 6 1136
NM_015133.4 NP_055948.2 6 1133
XM_005255190.2 XP_005255247.1 6 1136
XM_006720869.2 XP_006720932.1 6 2566
XM_011522429.2 XP_011520731.1 7 1160
XM_011522430.2 XP_011520732.1 7 1160
XM_011522431.2 XP_011520733.1 7 1160
XM_011522432.2 XP_011520734.1 7 1160
XM_011522433.2 XP_011520735.1 2 1783
XM_017023079.1 XP_016878568.1 6 1136
XM_017023080.1 XP_016878569.1 6 1136
XM_017023081.1 XP_016878570.1 5 608
GenBank mRNA AB028989.2 - 6 1096
BC137123.1 - 6 1048 9021120
BC137124.1 - 6 1048 9021121
BC144486.1 - 6 1051 9053013
BC150266.1 - 6 1096 9007222
CA776361.1 - 1 158
CN405915.1 - 5 570

View Full Product Details