Gene Transcripts

Gene Symbol
CAMTA1
Entrez Gene ID
23261
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
Chromosome Location
Chr. 1: 6785324 - 7769706 on Build GRCh38
UniGene ID
Hs.397705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015215.3 NP_056030.1 20 5070
XM_011541083.2 XP_011539385.1 21 4955
XM_011541084.2 XP_011539386.1 21 4955
XM_011541085.2 XP_011539387.1 22 5027
XM_011541086.2 XP_011539388.1 20 4934
XM_011541087.2 XP_011539389.1 20 4883
XM_011541088.2 XP_011539390.1 20 4885
XM_011541089.2 XP_011539391.1 21 4616
XM_011541090.2 XP_011539392.1 21 4616
XM_017000773.1 XP_016856262.1 22 5707
XM_017000774.1 XP_016856263.1 21 4955
XM_017000775.1 XP_016856264.1 19 4864
XM_017000776.1 XP_016856265.1 23 5087
XM_017000777.1 XP_016856266.1 20 4595
XM_017000778.1 XP_016856267.1 20 4595
XM_017000779.1 XP_016856268.1 20 4546
GenBank mRNA AB020640.3 - 20 5070
AF111804.1 - 4 560
AK302088.1 - 13 2064
AK309727.1 - 10 2077
AY349360.1 - 4 555
BC051341.1 - 9 1754
BC151835.1 - 20 5070 8860115

View Full Product Details