Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015227.4 | NP_056042.1 | 3 | 511 | |
NM_133635.4 | NP_598368.2 | 3 | 511 | ||
NR_004858.1 | 3 | 511 | |||
XM_006723981.3 | XP_006724044.1 | 4 | 584 | ||
XM_011529504.2 | XP_011527806.1 | 4 | 585 | ||
XM_011529505.2 | XP_011527807.1 | 4 | 586 | ||
XM_011529506.2 | XP_011527808.1 | 3 | 524 | ||
XM_011529507.2 | XP_011527809.1 | 4 | 585 | ||
XM_011529508.2 | XP_011527810.1 | 4 | 586 | ||
XM_011529509.2 | XP_011527811.1 | 4 | 587 | ||
XM_011529510.2 | XP_011527812.1 | 4 | 494 | ||
XM_011529511.2 | XP_011527813.1 | 4 | 587 | ||
XM_011529512.2 | XP_011527814.1 | 4 | 587 | ||
XM_011529513.2 | XP_011527815.1 | 4 | 586 | ||
XM_017028306.1 | XP_016883795.1 | 3 | 524 | ||
XM_017028307.1 | XP_016883796.1 | 4 | 587 | ||
XM_017028308.1 | XP_016883797.1 | 4 | 587 | ||
GenBank mRNA | AB023175.1 | - | 3 | 500 | |
AJ302079.2 | - | 3 | 500 | ||
AJ302080.2 | - | 3 | 500 | ||
AJ575591.1 | - | 3 | 495 | ||
AK290732.1 | - | 3 | 508 | ||
AK298080.1 | - | 3 | 508 | ||
AL110285.1 | - | 3 | 497 | ||
AY066015.2 | - | 3 | 500 | ||
BC064623.1 | - | 3 | 493 | 5727996 | |
BI560842.1 | - | 3 | 513 | ||
BI669244.1 | - | 3 | 514 | ||
HG380510.1 | - | 4 | 551 |