Gene Transcripts

Gene Symbol
LRRC8B
Entrez Gene ID
23507
Gene Name
leucine rich repeat containing 8 family member B
Gene Aliases
TA-LRRP, TALRRP
Chromosome Location
Chr. 1: 89522627 - 89597861 on Build GRCh38
UniGene ID
Hs.482017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134476.1 NP_001127948.1 7 2041
NM_015350.2 NP_056165.1 5 2098
XM_005270701.4 XP_005270758.1 5 1967
XM_005270702.4 XP_005270759.1 6 1988
XM_005270703.4 XP_005270760.1 5 3735
XM_011541143.2 XP_011539445.1 6 2025
XM_011541144.2 XP_011539446.1 7 2337
XM_011541145.2 XP_011539447.1 8 2362
XM_011541146.2 XP_011539448.1 6 3792
XM_011541148.2 XP_011539450.1 6 2013
XM_017000883.1 XP_016856372.1 6 2987
XM_017000884.1 XP_016856373.1 5 3889
XM_017000885.1 XP_016856374.1 5 2059
XM_017000886.1 XP_016856375.1 5 1963
GenBank mRNA AF385436.1 - 5 2098
AY358112.1 - 4 1849
AY640232.1 - 3 1679
BC030607.2 - 7 2277 4824652
BC053565.1 - 1 731
BC060782.1 - 7 1983 5298560
D86984.2 - 5 2404

View Full Product Details