Gene Transcripts
- Gene Symbol
- HEBP2
- Entrez Gene ID
- 23593
- Gene Name
- heme binding protein 2
- Gene Aliases
- C6ORF34B, C6orf34, PP23, SOUL
- Chromosome Location
- Chr. 6: 138404199 - 138415593 on Build GRCh38
- UniGene ID
- Hs.486589
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001326380.1 |
NP_001313309.1
|
1, 2 |
179 |
|
NM_001326381.1 |
NP_001313310.1
|
1, 2 |
410 |
|
NM_014320.2 |
NP_055135.1
|
1, 2 |
410 |
|
XM_017010631.1 |
XP_016866120.1
|
1, 2 |
383 |
|
GenBank mRNA |
AF117616.1 |
- |
1, 2 |
388 |
|
AF411610.1 |
- |
2 |
50 |
|
AK057928.1 |
- |
1, 2 |
325 |
|
AY427823.1 |
- |
1, 2 |
292 |
|
BC008205.1 |
- |
1, 2 |
274 |
3869026 |
BC010290.1 |
- |
1, 2 |
287 |
3445763 |
BC016806.1 |
- |
1, 2 |
390 |
|
BC016807.1 |
- |
1, 2 |
390 |
|
BC093037.1 |
- |
1, 2 |
273 |
30333215 |
BF974311.1 |
- |
|
410 |
|
View Full Product Details