Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014521.2 | NP_055336.1 | 3 | 469 | |
XM_011510891.2 | XP_011509193.1 | 3 | 489 | ||
XM_011510892.1 | XP_011509194.1 | 3 | 478 | ||
XM_011510893.1 | XP_011509195.1 | 3 | 403 | ||
XM_011510894.2 | XP_011509196.1 | 3 | 2902 | ||
GenBank mRNA | AF147747.1 | - | 3 | 411 | |
AK291209.1 | - | 3 | 436 | ||
BC057396.1 | - | 3 | 480 | 6138465 | |
BX647198.1 | - | 3 | 396 | ||
BX648877.1 | - | 3 | 584 | ||
DQ232895.1 | - | 3 | 443 |