Gene Transcripts

Gene Symbol
KBTBD2
Entrez Gene ID
25948
Gene Name
kelch repeat and BTB domain containing 2
Gene Aliases
BKLHD1
Chromosome Location
Chr. 7: 32868166 - 32892144 on Build GRCh38
UniGene ID
Hs.372541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015483.2 NP_056298.2 4 1258
XM_005249696.2 XP_005249753.1 4 1114
XM_005249697.2 XP_005249754.1 4 847
XM_011515256.2 XP_011513558.1 4 993
XM_011515257.2 XP_011513559.1 4 727
GenBank mRNA AB040922.1 - 1 1882
AF151831.1 - 3 667
AF161402.1 - 1 21
AK024557.1 - 1 223
AK292802.1 - 4 980
BC037887.2 - 4 1162 5274706
BC047107.1 - 4 995 5275217

View Full Product Details