Gene Transcripts
- Gene Symbol
- FBXO8
- Entrez Gene ID
- 26269
- Gene Name
- F-box protein 8
- Gene Aliases
- DC10, FBS, FBX8
- Chromosome Location
- Chr. 4: 174236658 - 174284251 on Build GRCh38
- UniGene ID
- Hs.76917
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012180.2 |
NP_036312.2
|
4, 5 |
1415 |
|
XM_017008020.1 |
XP_016863509.1
|
4, 5 |
679 |
|
GenBank mRNA |
AF115514.1 |
- |
4, 5 |
799 |
|
AF174596.1 |
- |
4, 5 |
631 |
|
AF201932.1 |
- |
4, 5 |
824 |
|
AF233224.1 |
- |
4, 5 |
670 |
|
AK026319.1 |
- |
4, 5 |
838 |
|
AK123740.1 |
- |
4, 5 |
1428 |
|
AK314483.1 |
- |
4, 5 |
833 |
|
AY358721.1 |
- |
4, 5 |
801 |
|
BC014679.2 |
- |
4, 5 |
817 |
3857760 |
BC014903.1 |
- |
4, 5 |
1415 |
|
BC040456.1 |
- |
5, 6 |
708 |
4281341 |
KU178588.1 |
- |
3, 4 |
565 |
|
View Full Product Details